Would you like to log in again?
CORIO™ CP Heating Circulator Baths, JULABO USA, Inc.
Supplier: Julabo
CORIO™
9013506.33 9013506.33 9013506.22 9013526.33 9013504.33 9013504.22 9013512.22 9013512.33 9013526.33 9013506.22 9013526.22 9013512.22 9013504.22 9013512.33 9013526.22 9013504.33
76353-826EA 5225.29 USD InStock
CA76353-828 76353-826 CA76353-826 CA76353-834 76353-828 CA76353-824 76353-822 CA76353-832 76353-832 CA76353-822 76353-824 CA76353-830 CA76353-820 76353-834 76353-820 76353-830
CORIO™ CP Heating Circulator Baths, JULABO USA, Inc.
Circulating Thermostats Open Circulator Systems Baths with Integral Circulators
CORIO™ CP heating circulator combinations with stainless steel bath tanks for internal or external temperature applications from 20 to 200 °C.
  • Infinitely adjustable pressure pump
  • Easy to switch between internal and external circulation
  • Bright LED display
  • USB and RS232 communication ports
For internal and external temperature applications. High quality stainless steel bath tanks with bath lid and drain tap.
CORIO CP circulators feature bright LED display with push button control. A lever below the display enables simple adjustment between internal and external fluid flow. The USB and RS232 ports facilitate external communication with PC or PLC systems. Pump connections: M16×1 male threads.
To adapt to your individual application there is a comprehensive range of accessories.
Certifications: CE certified.
Delivery information: Includes CORIO CP immersion circulator, mounting bridge, bath tank, bath tank lid, 8 and 12 mm hose barbs, integrated cooling coil and drain tap. Supplied without accessories.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty