The classically designed VWR® four-leg lab bench is the ideal basic bench, the simplest and the most cost-effective option in range of workbenches.
- Steel table frame, epoxy powder coated light neutral
- Supports up to 2000 lbs.
- Easily add a variety of accessories to create a complete work space
- Height adjustable with an allen key between 27.56 to 40.55"
- Phenolic and laminate worksurfaces available
Frame color is light neutral. Top features black phenolic or black laminate options. Laminate tops are 1.1875" thick, phenolic tops are 0.75" thick.
Accessories information: Additional accessories are available.
Delivery information: Assembly is required. Tables are delivered unassembled, unless otherwise ordered.