Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® 4-Leg Lab Benches

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
14-CS936 14-CS944 14-CS908 14-CS915 14-CS941 14-CS927 14-CS904 14-CS924 14-CS506-VWR CS718-VWR 14-CS909 14-CS933 14-CS505-VWR 14-CS911 14-CS942 14-CS913 14-CS940 14-CS503-VWR 14-CS919 14-CS945 14-CS502-VWR 14-CS934 14-CS905 14-CS912 14-CS916 14-CS907 14-CS948 14-CS918 14-CS926 14-CS901 14-CS914 14-CS910 14-CS931 CS715-VWR 14-CS923 14-CS501-VWR 14-CS946 14-CS906 14-CS504-VWR 14-CS947 CS712-VWR 14-CS920 14-CS921 14-CS925 14-CS902 14-CS932 14-CS929 14-CS903 14-CS943 14-CS917 14-CS949 14-CS928 14-CS935 14-CS930 14-CS922
76355-628EA 2091.08 USD InStock
76355-628 76355-626 76906-188 76906-186 76906-192 76355-590 76906-194 76355-594 76906-190 76355-592 76355-630 76355-598 76355-554 76355-596 76355-552 76355-634 76355-632 76355-556 76355-638 76355-636 76355-642 76355-640 76355-568 76355-602 76355-646 76355-644 76355-600 76355-606 76355-648 76355-604 76355-608 76355-572 76355-570 76355-652 76355-576 76355-650 76355-574 76355-656 76355-612 76355-654 76355-610 76355-578 76355-616 76355-614 76355-658 76355-618 76906-196 76355-582 76355-580 76355-586 76355-620 76355-584 76355-624 76355-588 76355-622
VWR® 4-Leg Lab Benches
Furniture Tables and Workbenches
The classically designed VWR® four-leg lab bench is the ideal basic bench, the simplest and the most cost-effective option in range of workbenches.

  • Steel table frame, epoxy powder coated light neutral
  • Supports up to 2000 lbs.
  • Easily add a variety of accessories to create a complete work space
  • Height adjustable with an allen key between 27.56 to 40.55"
  • Phenolic and laminate worksurfaces available

Frame color is light neutral. Top features black phenolic or black laminate options. Laminate tops are 1.1875" thick, phenolic tops are 0.75" thick.

Accessories information: Additional accessories are available.

Delivery information: Assembly is required. Tables are delivered unassembled, unless otherwise ordered.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty