Would you like to log in again?
COLUMN HITRAP MABSELECT SURE LX 5X5 ML
Supplier: Cytiva
Column Hitrap Mabselect Sure 5X5 Ml^ HiTrap MabSelect SuRe LX Column, for purification of monoclonal antibodies from large sample volumes, novel alkali-tolerant recombinant Protein A ligand allow cleaning-in-place, size: 5x5ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN HITRAP MABSELECT SURE LX 5X5 ML 76355-680 29157185 Each(25ml) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty