Would you like to log in again?
FLASK ONLY BALL SPINNER GLASS 100 ML
Supplier: Bellco Glass
Flask only Ball Spinner Glass 100 Ml^ Ball Spinner Flask only, Material: Type 1 Class A Borosilicate Glass, Center Cap Size: 70mm, Sidearm Size: 32mm, Glass ball houses a magnet for gentle agitation of micro-carrier beads, bottom indent prevents accumulation under the impeller, Size: 100 ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FLASK ONLY BALL SPINNER GLASS 100 ML 76356-728 1965-94010 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty