Would you like to log in again?
HOMOGENIZER CONICAL BOROSILCATE GLS 50ML
Supplier: Bellco Glass
Homogenizer Conical Borosilcate Gls 50Ml^ Conical Homogenizer, Material: Borosilicate Glass, designed for use with homogenizing motors, ideal for tissues such as muscle, heart and lung, has a large area for most efficient grinding, Capacity: 50 mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HOMOGENIZER CONICAL BOROSILCATE GLS 50ML 76356-854 1976-10025 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty