Would you like to log in again?
KAPUTS POLYPROPYLENE BLACK 20 MM CS500
Supplier: Bellco Glass
Kaput Polypropylene Black 20 Mm CS500^ Kaput, Non-Toxic Autoclavable Polypropylene, Ideal for culture tubes and flasks with DeLong style neck and access ports, Color: Black, stainless steel closures, Weight: 160 oz, Size: 20mm, Dimensions: 16 x 12 x 8 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KAPUTS POLYPROPYLENE BLACK 20 MM CS500 76357-042 2007-20001 Case of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty