Would you like to log in again?
FLASK SHAKE 3SIDE BAFFLE 38MM 500ML CS12
Supplier: Bellco Glass
Flask Shake 3Side Baffle 38Mm 500Ml CS12^ Shake Flask, with 3 Side Baffles DeLong Style Neck, material: Type 1 Class A Borosilicate Glass, are ideal for preliminary fermentation and bacterial growth studies, basic tool for growth studies of aerobic microbial cultures, Closure Size: 38mm, size: 500ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FLASK SHAKE 3SIDE BAFFLE 38MM 500ML CS12 76357-424 2540-00500 Case of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty