Would you like to log in again?
BEAKER GRIFFIN HEAVY WALL 400ML CS12
Supplier: Bellco Glass
Beaker Griffin Heavy Wall 400Ml CS12^ Griffin Beaker, Heavy Wall, Type 1 Class A Borosilicate Glass, with a low coefficient of expansion, meet ASTM-960 type 1 specifications for classification design capacity and markings, white permanent graduations, capacity: 400 ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BEAKER GRIFFIN HEAVY WALL 400ML CS12 76357-628 5620-00400 Case of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty