Would you like to log in again?
CAP SCREW ROUX CULT BOTTLE 1200 ML CS2
Supplier: Bellco Glass
Cap Screw Roux Cult Bottle 1200 Ml CS2^ Screw Cap, Roux Cult Bottle, Ideal for large area mono-layer cell culture growth, Schott Blank glass, Offset neck prevents culture from entering neck area, Working volume is approximately one half total volume, Dimensions: 132x56x280mm, Size: 1200ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAP SCREW ROUX CULT BOTTLE 1200 ML CS2 76357-640 5632-12000 Case of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty