Would you like to log in again?
LINER PTFE SILICONE 32MM W/PP CAP CS10
Supplier: Bellco Glass
Liner PTFE Silicone 32Mm W/Pp Cap CS10^ PTFE Silicone Liner, Use with open top Polypropylene caps for applications requiring addition withdraw of vessel contents by syringe, Autoclavable to 140C, size: 32 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LINER PTFE SILICONE 32MM W/PP CAP CS10 76357-668 5637-00038 Case of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty