Would you like to log in again?
Somatic Cell Count Microscope Slide
Supplier: Bellco Glass
5638-01930
76357-804CS 385.68 USD InStock
76357-804
Somatic Cell Count Microscope Slide
Slides Microscope Slides
The somatic Cell Count Slide is useful for direct microscopic counts of somatic cells in cows milk.
Spread 0.01 ml sample onto each circle than dry and stain. Count cells under oil immersion microscopy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty