Would you like to log in again?
MAGNETIC STIRRER 1 POSITION 115V 8 L
Supplier: Bellco Glass
Magnetic Stirrer 1 Position 115V 8 L^ Bell-Stirr Magnetic Stirrer, 1 Position 115v, designed for growing both mammalian and bacterial cultures where minimal heat transfer from the magnetic stirrer to the culture vessel is essential, Max Flask Size: 8 Liter
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MAGNETIC STIRRER 1 POSITION 115V 8 L 76358-026 7760-06003 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty