Would you like to log in again?
COMPRESSION FITTING ASBY 1/8 F/10MM PORT
Supplier: Bellco Glass
Compression Fitting Asby 1/8 F/10Mm Port^ Compression Fitting Assembly 1/8 for 10mm Port, compression fitting with 1/8in pipe included, Fits into 10mm port and a 3L metric headplate. 316l Stainless Steel, Size: 1/8in Pipe, Fits 10mm Port & 3l Metric Headplate
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COMPRESSION FITTING ASBY 1/8 F/10MM PORT 76358-304 7909-00018 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty