Would you like to log in again?
VWR® Wire Racks
Supplier: VWR
VWR®
120765V 120760V 120761V 120772V 120089V 120766V 120770V 120767V 120759V HS23072V 120763V 120768V 120769V 120764V 120771V
76358-666CS 1153.53 USD InStock
76358-666 76358-656 76358-678 76358-668 76358-658 76358-662 76358-652 76358-674 76358-664 76358-676 76358-654 76358-680 76358-670 76358-660 76358-672
VWR® Wire Racks
Racks Tube Racks
These HDPE coated wire racks are durable for transport or sample handling.
  • Chemically-resistant
  • Racks can be used between temperatures of –40 °C and 121 °C
  • Slotted bottoms
  • Holds all brands and types of tubes/bottles for each size range
  • Square wells and three deck levels help keep tubes secure
  • Autoclavable
Suitable for conical, round or flat-bottom glass or plastic tubes and bottles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty