Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Avantor Single-Use Bioprocessing Bags, Avantor Fluid Handling

Supplier: Avantor Fluid Handling
1002531 1002527 1002502 1002535 1002537 1002501 1002530 1002528 1002534 1002504 1002532 1002536 1002526 1002529 1002503 1002538 1002533
76358-708CS 1386.76 USD InStock
76358-708 76358-714 76358-704 76358-706 76358-710 76358-688 76358-700 76358-712 76358-702 76358-684 76358-696 76358-686 76358-698 76358-692 76358-682 76358-694 76358-690
Avantor Single-Use Bioprocessing Bags, Avantor Fluid Handling
Bags Media Bags
Avantor single-use bioprocessing bags are designed to transfer and store your products throughout the drug-manufacturing process. Made from proprietary FlexFilm™ material – a thick, co-extruded film featuring polyethylene inner and outer layers that provide extra resistance to a wide range of chemicals – Avantor single-use bags are provided in a range of standard sizes to meet the needs of many bioprocessing applications.

  • Bag film made from proprietary ULDPE with high clarity and flexibility
  • Standard 2D bag assemblies provided at volumes ranging from 50 ml to 50 L
  • Standard 3D bag chambers of 100, 200, and 500 L for faster custom assembly deliveries
  • Custom 3D bag chamber volumes up to 3000 L for large volume processes
  • Bag assemblies provided irradiated with SAL 10⁻⁶ and USP Class VI certification

Avantor's standard 2D bag assemblies are available with C-Flex tubing with plugged ends for welding and silicone tubing with either luer or MPC connectors for easy operator connections. The custom bag assemblies, both 2D and 3D, allow for user-specified tubing materials and connection types including a range of sterile connectors from all major manufacturers.

The 2D bag assemblies are designed for hanging which allows ease of use and complete fluid recovery. The standard 3D bag chambers available at 100, 200, and 500 L are top-ported for easy use in a variety of totes and container configurations.

Delivery information: P1 and P2 are supplied with clamps.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty