Would you like to log in again?
ZymoBIOMICS Lysis Solution, Zymo Research
Supplier: Zymo Research
D4300-1-150
76358-806EA 136 USD InStock
76358-806
ZymoBIOMICS Lysis Solution, Zymo Research
Buffers Extraction Buffers
ZymoBIOMICS Lysis Solution is designed to be used with the ZymoBIOMICS DNA extraction kits. Used in combination with the ZR BashingBead Lysis Tubes, it assists with microbial lysis. It is certified to be low-bioburden to ensure minimal background contaminants for low abundance microbiome samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty