Would you like to log in again?
ZYMO-SPIN 384 WELL PLATE PP W/1.2UM MEMB
Supplier: Zymo Research
Zymo-Spin 384 Well Plate PP W/1.2Um Memb^ Zymo-Spin 384 Well Plate, Material: Polypropylene spin plate with 1.2 um glass fiber membrane. 140 ul/well, long drip, 2 Pack
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ZYMO-SPIN 384 WELL PLATE PP W/1.2UM MEMB 76358-918 C2012 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty