Would you like to log in again?
PIG® HazMat Spill Kit in 20-Gallon High-Visibility Container, NEW PIG
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PIG®
KIT312
76358-922EA 422.83 USD InStock
76358-922
PIG® HazMat Spill Kit in 20-Gallon High-Visibility Container, NEW PIG
Spill Control Materials
Hi-viz container is easily seen even in low light or from a distance to help make spill response even faster.
  • PIG HazMat Socks stop spreading spills; PIG HazMat Pads and Pillows absorb quickly
  • Prepacked containers are organized for easy access
  • Lime green drum is easy to locate
  • Lightweight, polypropylene drum resists chemicals and keeps contents clean and dry
  • Bold "Spill Kit" lettering for quick identification
  • Tamperproof seal helps prevent pilfering of spill response supplies
PIG® HazMat absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide.
Nonthreaded lid lifts off for easy access (not rated as a shipping container).
Only PIG® Spill kits feature PIG® Absorbents proven for rapid response.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty