Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Chemotherapy Waste Bags, Medegen Medical Products
Supplier: Medegen Medical Products
47-CBL4 9230 4210 9233
76358-992CS 131.04 USD InStock
76358-992 76359-014 76359-100 76359-012
Chemotherapy Waste Bags, Medegen Medical Products
Bags Waste Disposal Bags
Chemotherapy waste collection bags are ideal for transport of anti-neoplastic medications and chemotherapy waste.
  • Heavy-duty construction for puncture and tear resistance
  • Dual tested for use in all 50 states
  • Made in the USA
Bags feature reinforced seals for leak resistance and meet all labeling requirements for CYTA collection. All bags are color-coded (yellow film) and feature bold labeling with International Biohazard Symbol for easy identification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty