Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Water Soluble Laundry Bags, Medegen Medical Products
Supplier: Medegen Medical Products
1-347 547-A 1-341 1-342 1-348 548-A 1-344 541-A 1-345
76359-142CS 147.87 USD InStock
76359-142 76359-032 76359-140 76359-042 76359-036 76359-034 76359-144 76359-038 76359-040
Water Soluble Laundry Bags, Medegen Medical Products
Bags Laundry Bags
These laundry bags are designed for the safe collection and handling of potentially contaminated or infectious linen and clothing.
  • 100% Biodegradable polyvinyl alcohol fill dissolves completely into a non-toxic aqueous solution
  • Allows for easy isolation of contaminated and heavily soiled linens to reduce risks of cross-contamination
  • Superior wet strength holdout capacity and order control
  • Hot water bags dissolve at 150 °F
  • Cold water bags dissolve at 55 °F
These bags are impervious to bacteria and viruses. They can be placed directly into the washing machine, eliminating the need for staff to come in direct contact with the contents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty