Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Dual-Tested Autoclave Biohazardous Waste Bags, Medegen Medical Products
Supplier: Medegen Medical Products
AC1923C AC1419C AC3847C AC2530C AC1419R AC2530R AC2535C AC2535R AC3847R AC3138R AC3138C AC1923R
76359-164CS 234.03 USD InStock
76359-164 76359-154 76359-162 76359-152 76359-174 76359-168 76359-158 76359-166 76359-156 76359-160 76359-172 76359-170
Dual-Tested Autoclave Biohazardous Waste Bags, Medegen Medical Products
Bags Waste Disposal Bags
Dual tested autoclave bags are built to withstand the rigors of steam sterilization while meeting the essential requirements for the collection, storage, transport, and processing of autoclavable biohazardous waste.
  • Biohazardous labeling in English and Spanish
  • Twist ties included for easy closure
  • Made in the USA
Polypropylene film has an operating temperature of 274 °F (134 °C).
Certifications: Bags meet both ASTM D1922 (Elmendorf Tear 480g) and ASTM D1709 Dart Impact (160g) requirements. They comply with US DOT 40CFR Sections 173 -197 and regulations in all 50 states including CA, FL, and PA.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty