Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Biohazardous Waste Bags, Medegen Medical Products
Supplier: Medegen Medical Products
F135 F110 2635 F117 F118 37-97 2634 F124 47-41 F149 F169 47-43 F145 45-75 RD650 45-43 47-71 45-30 50-40 45-50 F130 RD630 47-45 45-54 F109 45-45 RD610 F116 50-42
76359-186CS 133.06 USD InStock
76359-186 76359-066 76359-184 76359-202 76359-188 76359-200 76359-080 76359-182 76359-084 76359-180 76359-082 76358-984 76359-076 76359-098 76359-198 76359-074 76359-196 76359-078 76359-178 76359-090 76359-190 76359-094 76358-986 76359-194 76359-092 76359-192 76359-128 76359-126 76359-204
Biohazardous Waste Bags, Medegen Medical Products
Bags Waste Disposal Bags
These biohazardous waste bags are manufactured to meet the specific performance standards in accordance with Federal and specific State Guidelines.
  • Biohazardous labeling in both English and Spanish
  • Latex-free and environmentally safe for incineration disposal
Premium LLDPE liners ensure consistent strength and superior performance.
Certifications: Bags meet both ASTM D1922 (Elmendorf Tear 480g) and ASTM D1709 Dart Impact (160g) requirements. Complies with US DOT interstate transport of biohazardous waste and all 50 states.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty