Would you like to log in again?
VWR® Nitrile Chemical Safe Gloves
Supplier: VWR
VWR®
GNF15130GRNLG GNF15130GRNXL GNF15130GRNMD GNF15130GRNSM GNF15130GRN2X
76359-828CS 454.51 USD InStock
76359-828 76359-822 76359-820 76359-826 76359-824
VWR® Nitrile Chemical Safe Gloves
Gloves
VWR® Nitrile Chemical Safe Gloves are specially formulated with a nitrile compound that provides outstanding protection against a wide range of chemicals, including most solvents and acids, strong detergents, greases, oils, and animal fats.
  • 15 mil (0.38 mm) thickness
  • 13" (330 mm) length
  • Fully textured
  • Flock lined
  • Exceptional flexibility and comfortable to wear
Patterned palm for excellent grip in both wet and dry conditions. Resistance against abrasion, cuts, snags, and puncture, ensures longer wear and better protection than natural rubber gloves.
Certifications: Gloves comply to European standards EN388, EN ISO 374-1 Type A & EN ISO 374-5. Certified to CE category 3 for complex risks.
Packaging: Gloves are packaged 12 pairs per bag, and 144 pairs per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty