Would you like to log in again?
Brady® BatteryBlock™ Battery Cable Lockout - Commercial Vehicles, Brady Worldwide
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
150822 150821 150823 150820
76359-906EA 26 USD InStock
76359-906 76359-908 76359-910 76359-912
Brady® BatteryBlock™ Battery Cable Lockout - Commercial Vehicles, Brady Worldwide
Security Locks
Get easy prevention of an accidental battery connection during service projects on commercial vehicles with the BatteryBlock™ Battery Cable Lockout.
  • A tool that fits virtually any space
  • Low profile design means it easily fits into tight working areas
  • Small size fits endline battery cables
  • Keep it locked out with space for two safety padlocks (sold separetely)
Innovative non-conductive post slips through the terminal end of the battery cable to prevent it from slipping or being pulled out of the device.
Certifications: OSHA 29 CFR 1910.147.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty