Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Brady® BatteryBlock™ Power Connector Lockout - Forklift, Brady Worldwide

Supplier: Brady

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

150841
76359-914EA 35.96 USD InStock
76359-914
Brady® BatteryBlock™ Power Connector Lockout - Forklift, Brady Worldwide
Security Locks
For forklift safety, proper operation is a main concern, but a secure lockout can be just as important. Keep forklifts and lift vehicles locked out and locked down with the BatteryBlock™ power connector lockout.

  • Device fits on 175 A and 350 A power connectors with and without handles
  • Single-piece design is easy to store and manage so users don't have to worry about losing pieces
  • Manage use and safety by applying two padlocks from authorized employees

Non-conductive pins ensures the device stays secure during maintenance projects and off-shift periods.

Certifications: OSHA 29 CFR 1910.147.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty