Would you like to log in again?
Brady® Safety Inspection Timers, Brady Worldwide
Supplier: Brady
150744 150741 150743 150742 150740
76360-010PK 324.47 USD InStock
76360-010 76360-012 76360-014 76360-006 76360-008
Brady® Safety Inspection Timers, Brady Worldwide
Timers Digital Timers
Shed some light on inspections by supplementing inspection tags with the inspection timer, and get an inspection status at a glance.
  • Review and reset; after completing an inspection, reset the timer so it is ready for the next inspection
  • Perfect for facilities that have internal audit and inspection schedules
  • Sturdy ABS plastic construction
Unlike tags, the bright color LED lights provide an inspection status from a distance and break through the grime of any facility
IP 65 rated timers will survive harsh industrial environments, including exposure to water, dust, extreme temperatures and more.
Safety inspection timers are ideal for use in facilities that have multiple eyewash stations, emergency showers, emergency exit doors, exit signs, fire extinguishers, or first aid kits.
Certifications: Ingress Protection Rating IP 65.
Delivery information: All timers are supplied with three AAA Alkaline batteries.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty