Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Cole-Parmer® elements™ Magnetic Stirrers, Cole-Palmer
Supplier: Cole Parmer
84003-82 84003-80 84003-81
76360-044EA 186.43 USD InStock
76360-044 76360-040 76360-042
Cole-Parmer® elements™ Magnetic Stirrers, Cole-Palmer
Stirrers Magnetic Hotplate Stirrers, Multi-position
This unique multiposition stirrer allows the user to decide how many stirring plates to attach (up to 10) based on mixing needs.
  • Requires minimal work space
  • Simple touch and slide operation
  • LCD display with real time parameter
  • Corrosion-resistant glass stirring top
  • Liquid proof USB attachement
  • Cables can be easily removed or attached to the stir plate
By means of either direct USB connection or via the included cables it is simple to attach or remove a stirring plate and configure to suit the work area. Each stir plate edge lights up to allow accurate positioning of the vessel. Multiple stirring options allow a choice between one direction or reversing every 45 seconds for thorough mixing. Built in timer can be set for up to 9 hours or continuous operation.
Certifications: IP protection class: IP 44
Delivery information: Kit includes control module, four stirring plate modules, USB cords, power cord and AC adapter.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty