Would you like to log in again?
GAS CYLINDER CALIBRATION O2 IN N2 20.8%
Supplier: ORS NASCO, INC.
Gas Cylinder Calibration O2 In N2 20.8%^ Calibration Gas Cylinder, Oxygen in Nitrogen, 20.8%, Designed for portable gas detectors, Quick and convenient way to check response of equipment, Lightweight carrying case, Material: Aluminum, Economically priced
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GAS CYLINDER CALIBRATION O2 IN N2 20.8% 76360-248 454-10028028 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty