Would you like to log in again?
Diamond® RealSeal™ Bottles, Wide Mouth, High Density Polyethylene, Globe Scientific
Supplier: Globe Scientific
7011000 7010250 7010125 7010060 7174000 7010500 7184000 7012000 7010030
76360-274EA 51.65 USD InStock
76360-274 76360-264 76360-266 76360-268 76360-258 76360-270 76360-260 76360-272 76360-262
Diamond® RealSeal™ Bottles, Wide Mouth, High Density Polyethylene, Globe Scientific
Bottles Plastic Bottles
Diamond® RealSeal™ Plastic Laboratory Bottles meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.
  • Safe for freezer storage down to –100 °C
  • Resistant to most corrosives
  • Medical grade resins meet demanding requirements
  • Precision molded for consistent wall thickness and smooth surfaces
  • Leakproof, linerless cap for secure closing
  • Laboratory bottle quality and performance with savings associated with economy bottles
  • Safe, shatter-resistant alternative to glass bottles
These are available in a wide range of shapes, sizes, and materials to accomodate most application needs. Small or large, wide mouth or narrow, natural or amber, even large format with handles, the Diamond® RealSeal™ line includes the most frequently used and less common bottles.
These wide mouth bottles are perfect for sample collection and facilitate rapid filling and emptying of solids, powders, and viscous liquids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty