Would you like to log in again?
Diamond® RealSeal™ Bottles, Amber, Wide Mouth, High Density Polyethylene, Globe Scientific
Supplier: Globe Scientific
7011000AM 7010060AM 7010250AM 7010500AM 7010030AM 7010125AM 7012000AM
76360-320EA 40.56 USD InStock
76360-320 76360-310 76360-312 76360-314 76360-316 76360-318 76360-308
Diamond® RealSeal™ Bottles, Amber, Wide Mouth, High Density Polyethylene, Globe Scientific
Bottles Plastic Bottles
Diamond® RealSeal™ Plastic Laboratory Bottles from Globe Scientific meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.
  • Safe for freezer storage down to –100 °C
  • Resistant to most corrosives
  • Medical grade resins meet demanding requirements
  • Precision molded for consistent wall thickness and smooth surfaces
  • Leakproof, linerless cap for secure closing
  • Laboratory bottle quality and performance with savings associated with economy bottles
  • Safe, shatter-resistant alternative to glass bottles
Diamond® RealSeal™ Bottles are available in a wide range of shapes, sizes, and materials to accomodate most application needs. Small or large, wide mouth or narrow, natural or amber - even large format with handles - the Diamond® RealSeal™ line includes the most frequently used and less common bottles.
These wide mouth bottles are perfect for sample collection and facilitate rapid filling and emptying of solids, powders, and viscous liquids. Amber bottles reduce UV light transmission to protect light sensitive contents during storage and transport. All Diamond® RealSeal™ amber bottles comply with US Pharmacopoeia 23661 requirements for maximum light transmission.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty