Would you like to log in again?
BASE UNIT WITH 7INCH DRAWER 22X36X35IN
Supplier: EAGLE MHC
Base Unit With 7Inch Drawer 22X36X35In^ Base unit, standing height, (4) 7inch drawers with wire style pulls, Dimension: 22X36X35inch, Material: stainless steel construction, with adjustable intermediate shelf & removable back
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BASE UNIT WITH 7INCH DRAWER 22X36X35IN 76360-548 SSC-HDB7-36 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty