Would you like to log in again?
CABINET WALL W/CLOSED FRONT 13X18X36INCH
Supplier: EAGLE MHC
Cabinet Wall W/Closed Front 13X18X36Inch^ Wall cabinet, closed front, sloped top, Dimension: 13X18X36inch, right-hand hinged door with wire style pull, Material: stainless steel construction, (2) adjustable intermediate shelves
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CABINET WALL W/CLOSED FRONT 13X18X36INCH 76360-810 SSC-WCHRST1318 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty