Would you like to log in again?
CABINET TALL WIRE STYLE PULL 24X24X89IN
Supplier: EAGLE MHC
Cabinet Tall Wire Style Pull 24X24X89In^ Tall Cabinet, closed front, sloped top, Dimension: 24X24X89inch, left-hand hinged door with tempered glass insert and wire style pull, Material: stainless steel construction, (5) adjustable intermediate shelves
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CABINET TALL WIRE STYLE PULL 24X24X89IN 76360-976 SSCTCBGLST2424 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty