Would you like to log in again?
MICROPLATE CELLVIEW 96WELL BLACK ST CS16
Supplier: Greiner Bio-One
Microplate Cellview 96Well Black St CS16^ Cellview Microplate, 96 Well, Cycloolefin, F-Bottom (Chimney Well), Glass Bottom, Black, Advanced Tc, Lid, Sterile, Single Packed
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MICROPLATE CELLVIEW 96WELL BLACK ST CS16 76363-2408 655981 Case of 16 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty