Would you like to log in again?
QC Sets and Panels: Helix Elite™, Microbiologics
Supplier: Microbiologics
8206 8238
76367-708PK 346.84 USD InStock
76367-708 76367-710
QC Sets and Panels: Helix Elite™, Microbiologics
Assays Cellular Assays
All the required QC strains for the lab’s microbial detection/identification system, combined in one convenient panel.
  • One simple catalog number for fast, easy ordering
  • Protects the reputation by providing documentation and assurance that the laboratory can consistently produce accurate results
  • Ensures testing procedures, equipment and materials are working properly
  • Ready-to-use format saves time and money
  • Online certificate of analysis provides detailed strain information
  • Traceability to reference cultures ensures authenticity
  • Technical support experts available for guidance
Each instrument-specific QC set or panel contains the manufacturer recommended quality control strains for a particular instrument or test in an easy-to-use format. Each regulation-specific QC set or panel contains the regulation-specific (e.g. CLSI® or EUCAST) recommended quality control strains for a particular test in an easy-to-use format.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty