Would you like to log in again?
Dumont Style S5, Long, Brass, Dumont Switzerland
Supplier: Electron Microscopy Sciences
5306-S5-PO
76375-950EA 68.8 USD InStock
76375-950
Dumont Style S5, Long, Brass, Dumont Switzerland
Forceps
These are popular tweezers for handling and manipulating delicate parts. The brass is composed of Cu, Pb, Sn, Ni and Zn. Widely used for handling delicate components, especially in watch-making.
  • 00% antimagnetic brass alloy
  • High precision Swiss manufacturing
  • Perfectly finished to suit your application
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty