Would you like to log in again?
EMS Swiss Line Standard Tweezer Style 258 Peek, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
7801-258PEEK
76376-056EA 71.79 USD InStock
76376-056
EMS Swiss Line Standard Tweezer Style 258 Peek, Electron Microscopy Sciences
Forceps
Swiss made tweezers have tips with 30% carbon fibers. The tips are ESD safe, anti-magnetic, anti acid as well as temperature resistance up to 250 °C (short exposure up to 300 °C). Can be sterilized at 143 °C. Often used in the disc drive industry and for cer
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty