Would you like to log in again?
EMS Swiss Line Standard Tweezer Style 49D, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
7801-49D
76376-072EA 114.5 USD InStock
76376-072
EMS Swiss Line Standard Tweezer Style 49D, Electron Microscopy Sciences
Forceps
Swiss made standard tweezers have Delrin tips and are temperature resistant up to 115 °C. Delrin is a very soft material that is non scratching, nicking or marring. The tweezers are excellent for use in biotech applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty