Would you like to log in again?
EMS Swiss Line Cutting Tweezer Style 15AP, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
7802-15AP
76376-096EA 126.22 USD InStock
76376-096
EMS Swiss Line Cutting Tweezer Style 15AP, Electron Microscopy Sciences
Forceps
Swiss made tweezers are for cutting hair sprigs, electronic component wires, copper, gold, silver and other soft wires. The large head produces a parallel, strong cut.
  • Surface resistively is 10E12 ohm
  • Contamination free, high temperature 1000 °C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty