Would you like to log in again?
EMS Swiss Line Wafer Handling Tweezer Style ARG, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
7803-ARG
76376-110EA 112.71 USD InStock
76376-110
EMS Swiss Line Wafer Handling Tweezer Style ARG, Electron Microscopy Sciences
Forceps
Swiss made tweezers are designed for gripping wafers. These are for large size wafers, as well as for holding slides in the lab.
Dimensions: D: 3 mm, E: 3 mm, F: 3 mm, M: 1 mm, R: 52 mm, S: 42 mm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty