Would you like to log in again?
EMS Swiss Line Wafer Handling Tweezer Style NW, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
7803-NW
76376-112EA 61.4 USD InStock
76376-112
EMS Swiss Line Wafer Handling Tweezer Style NW, Electron Microscopy Sciences
Forceps
Swiss made tweezers are designed for gripping wafers.
Dimensions: M: 11 mm, R: 52 mm, S: 42 mm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty