Would you like to log in again?
EMS Cutting Tweezers Style 15A, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78325-06 78325-14A 78325-14AG 78325-11 78325-15AF 78325-04 78325-05 78325-03 78325-152 7802-15A
76376-156EA 95.73 USD InStock
76376-156 76376-180 76376-182 76376-094 76376-150 76376-174 76376-152 76376-166 76376-176 76376-154
EMS Cutting Tweezers Style 15A, Electron Microscopy Sciences
Forceps
Swiss made tweezers are for cutting hair sprigs, electronic component wires, copper, gold, silver and other soft wires. The large Large head produces a oblique strong cut at the cutter's tip. The cutting edge is limited to the tip area.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty