Would you like to log in again?
EMS High Precision Rubber Grip Cutting Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78325-07 78325-08 78325-10 78325-09
76376-158EA 123.78 USD InStock
76376-158 76376-160 76376-162 76376-164
EMS High Precision Rubber Grip Cutting Tweezers, Electron Microscopy Sciences
Forceps
Soft ESD safe ergonomic cushion grips (resistivity 10⁸ to 10⁹ Ohm) provide enhanced operator comfort and reduced finger stress when handling ESD sensitive components or small static items. Tips are carbon steel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty