Would you like to log in again?
EMS Biological Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78325-4SA 78325-3SA 78325-5SA 78325-7SA
76376-234EA 124.64 USD InStock
76376-234 76376-200 76376-222 76376-190
EMS Biological Tweezers, Electron Microscopy Sciences
Forceps
EMS biological grade tweezers are made of a superalloy that contains nickel, chromium, and molybdenum. The Swiss made tweezers are anti-acid, anti-magnetic, and have serrated handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty