Would you like to log in again?
EMS Mini CX Biological Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78325-M3E 78325-M5E
76376-256EA 90.2 USD InStock
76376-256 76376-264
EMS Mini CX Biological Tweezers, Electron Microscopy Sciences
Forceps
EMS biological grade tweezers are made of a superalloy (CX) that contains nickel, chromium, and molybdenum. The Swiss made tweezers are anti-acid, anti-magnetic.
  • Suitable to use under a microscope
  • Resist extreme temperatures, chemicals and other harsh conditions
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty