Would you like to log in again?
EMS Economy Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78326-48 78326-42 78326-53 78326-50 78326-46 78326-47 78326-49 78326-40 78326-54 78326-44 78326-43 78326-45 78326-51 78326-55 78326-52 78326-41
76376-278EA 25.83 USD InStock
76376-278 76376-300 76376-288 76376-302 76376-304 76376-292 76376-280 76376-294 76376-282 76376-274 76376-296 76376-284 76376-298 76376-276 76376-286 76376-290
EMS Economy Tweezers, Electron Microscopy Sciences
Forceps
Economy tweezers made of 304 stainless steel.
  • Inexpensive tweezers beneficial in non-critical applications
  • These tweezers are anti-acid/non-corrosive and anti-magnetic
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty