Quality tweezers are ideal for laboratory work. For precise handling transmission electron microscopy grid, tweezer styles #3, #3C, #4, #5, #51S, and 5B are recommended.
- Very popular tweezers
- Tweezers possess the highest quality
- These tweezers have an anti-glare satin finish
SA low carbon austenitic steel is non-magnetic and anti-acid. It is ideal for use where corrosion resistance and toughness are primary requirements.