Would you like to log in again?
EMS High End Medical Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78920-08 78920-01 78920-07 78920-03 78920-06 78920-09 78920-02 78920-04
76376-388EA 99.14 USD InStock
76376-388 76376-376 76376-378 76376-380 76376-382 76376-384 76376-386 76376-374
EMS High End Medical Tweezers, Electron Microscopy Sciences
Forceps
The medical tweezers have a high precision fine tip with perfect symmetry and balance. They are made of Inox steel with an electropolish finish and dot serrated handles for a perfect grip.
  • Can be sterilized
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty