Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Cryogenic Dot Label Set

Supplier: VWR
VWR®
VWR-C105
76376-440CS 8.71 USD InStock
76376-440
VWR® Cryogenic Dot Label Set
Labels
Cryogenic labels are made of a thermoplastic film that is coated with a glove-friendly cryogenic adhesive.

  • Will stick in temperatures ranging from 80 to 196 °C
  • Resistant to smearing

This special adhesive allows the label to remain strong and sticky so that it can be securely adhered to stainless steel, aluminum, painted surfaces, and powder-coated metal and iron.

Certifications: Meets ASTM standards. EN 342.

Delivery information: Each set contains an 0.375" dot and 0.5"Ø×1.28" strip.

Packaging: Roll contains 500 sets of each.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty